Learn More
Pyrantel Related Compound A Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Catalog No. 111003573 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
Catalog No. Quantity
11-100-3573 50 mg
1 options
Catalog No. 11-100-3573 Supplier MilliporeSigma Supelco Supplier No. PHR208150MG
Add to Cart
Add to Cart
Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards
TRUSTED_SUSTAINABILITY
Specifications
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Type Certified Reference Material
Quantity 50 mg
Grade Certified Reference Material