Learn More
U.S. Pharmacopeia Pyrantel Tartrate, 33401-94-4, MFCD00216022, 200mg
Supplier:  U.S. Pharmacopeia 1584105
Encompass
Molecular formula C11H14N2S.C4H6O6, Molecular weight 356.43, Melting Point 298.4 - 302 °F (148 - 150 °C), Synonyms: Pyrantel hydrogen tartrate
Catalog No. NC1503452