Learn More
Medchemexpress LLC Pyrantel tartrate | 33401-94-4 | 98.7% | 356.39 | 1 ML
SDP
Catalog No. 5000436243
Encompass_Preferred
Pyrantel tartrate is an orally active anthelmintic and an agonist of the nicotinic acetylcholine receptor (nAChR). It causes spasmodic muscle paralysis in parasites and is used in the study of parasitic infections.
  • Agonist of nicotinic acetylcholine receptor (nAChR)
  • Causes spasmodic muscle paralysis in parasites
  • Used in the study of parasitic infections such as ascariasis and hookworm infections
Catalog No. 50-004-36243 Supplier Medchemexpress LLC Supplier No. HY1264110MM/1ML
Pyrantel tartrate is an orally active anthelmintic and an agonist of the nicotinic acetylcholine receptor (nAChR). It causes spasmodic muscle paralysis in parasites and is used in the study of parasitic infections.
  • Agonist of nicotinic acetylcholine receptor (nAChR)
  • Causes spasmodic muscle paralysis in parasites
  • Used in the study of parasitic infections such as ascariasis and hookworm infections