Learn More
Medchemexpress LLC Pyrantel (tartrate) | 33401-94-4 | 98.7% | 356.39 | 500 MG
SDP
Catalog No. 5000365721
Encompass_Preferred
Pyrantel tartrate is an orally active anthelmintic and an agonist of the nicotinic acetylcholine receptor (nAChR). It induces spasmodic muscle paralysis in parasites, making it useful for studying parasitic infections like ascariasis, hookworm infections, intestinal worms (pinworm infections), and trichinosis.
  • Orally active anthelmintic
  • Agonist of the nicotinic acetylcholine receptor (nAChR)
  • Induces spasmodic muscle paralysis in parasites
  • Useful for studying various parasitic infections
Catalog No. 50-003-65721 Supplier Medchemexpress LLC Supplier No. 500MGHY12641
Pyrantel tartrate is an orally active anthelmintic and an agonist of the nicotinic acetylcholine receptor (nAChR). It induces spasmodic muscle paralysis in parasites, making it useful for studying parasitic infections like ascariasis, hookworm infections, intestinal worms (pinworm infections), and trichinosis.
  • Orally active anthelmintic
  • Agonist of the nicotinic acetylcholine receptor (nAChR)
  • Induces spasmodic muscle paralysis in parasites
  • Useful for studying various parasitic infections