Learn More
Apexbio Technology LLC PYRANTEL-TARTRATE-500MG
SDP
Catalog No. 5000569346
Encompass_Preferred
Pyrantel tartrate (CAS No 33401-94-4) is a compound classified under the category of antiparasitic agents primarily used in veterinary medicine It originates from tetrahydropyrimidines and functions as a cholinergic agonist targeting nicotinic acetylcholine receptors at the neuromuscular junction of nematodes This mechanism causes spastic paralysis in parasites facilitating their expulsion In vitro activity of pyrantel tartrate is generally observed in the nanomolar to low micromolar range depending on the organism It is commonly applied in studies involving cell models of nematode infections and is used in animal studies to evaluate its efficacy as an anthelmintic The compound is also utilized in drug screening efforts to discover new treatments for parasitic infections Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-69346 Supplier Apexbio Technology LLC Supplier No. C6718500
Pyrantel tartrate (CAS No 33401-94-4) is a compound classified under the category of antiparasitic agents primarily used in veterinary medicine It originates from tetrahydropyrimidines and functions as a cholinergic agonist targeting nicotinic acetylcholine receptors at the neuromuscular junction of nematodes This mechanism causes spastic paralysis in parasites facilitating their expulsion In vitro activity of pyrantel tartrate is generally observed in the nanomolar to low micromolar range depending on the organism It is commonly applied in studies involving cell models of nematode infections and is used in animal studies to evaluate its efficacy as an anthelmintic The compound is also utilized in drug screening efforts to discover new treatments for parasitic infections Experimental concentrations typically depend on the specific research design