Learn More
Cayman Chemical Pyrazinamide, Purity: ≥98%, CAS: 98-96-4, 10 g
SDP
Catalog No. 502985791
Encompass_Preferred
An antimycobacterial compound; inhibits the growth of the human M. tuberculosis H37Rv strain (MIC = 150 μg/ml); increases survival in M. tuberculosis H37Rv-infected mice when administered orally (0.1% w/w in the diet) or subcutaneously (2.5 mg single injection) on the day of infection; inhibits the growth of M. lepraemurium in infected mice
Catalog No. 50-298-5791 Supplier Cayman Chemical Supplier No. 2341610 G
An antimycobacterial compound; inhibits the growth of the human M. tuberculosis H37Rv strain (MIC = 150 μg/ml); increases survival in M. tuberculosis H37Rv-infected mice when administered orally (0.1% w/w in the diet) or subcutaneously (2.5 mg single injection) on the day of infection; inhibits the growth of M. lepraemurium in infected mice