Learn More
Medchemexpress LLC Pyrazinamide | 98-96-4 | MFCD00006325 | ≥98% | 500 MG
SDP
Catalog No. 5000408578
Encompass_Preferred
Pyrazinamide is a potent and orally active antitubercular antibiotic. It is a proagent that is converted to the active form pyrazinoic acid (POA) by PZase/nicotinamidase encoded by the pncA gene in M. tuberculosis.
Catalog No. 50-004-08578 Supplier Medchemexpress LLC Supplier No. HYB0271500MG
Pyrazinamide is a potent and orally active antitubercular antibiotic. It is a proagent that is converted to the active form pyrazinoic acid (POA) by PZase/nicotinamidase encoded by the pncA gene in M. tuberculosis.