Learn More
Medchemexpress LLC Pyrazinamide-d3 | 1432059-16-9 | 99.9% | 1 MG
SDP
Catalog No. 5000408506
Encompass_Preferred
Pyrazinamide-d3 is a deuterium-labeled variant of Pyrazinamide, a potent and orally active antitubercular antibiotic. It functions as a proagent, converting into the active form, pyrazinoic acid (POA), through the action of PZase/nicotinamidase in *M. tuberculosis*. The incorporation of deuterium has garnered interest for its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Potent and orally active antitubercular antibiotic
  • Acts as a proagent converted to active form pyrazinoic acid (POA)
Catalog No. 50-004-08506 Supplier Medchemexpress LLC Supplier No. HYB0271S1MG
Pyrazinamide-d3 is a deuterium-labeled variant of Pyrazinamide, a potent and orally active antitubercular antibiotic. It functions as a proagent, converting into the active form, pyrazinoic acid (POA), through the action of PZase/nicotinamidase in *M. tuberculosis*. The incorporation of deuterium has garnered interest for its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Potent and orally active antitubercular antibiotic
  • Acts as a proagent converted to active form pyrazinoic acid (POA)