Learn More
Apexbio Technology LLC PYRAZINAMIDE-D3-1MG
SDP
Catalog No. 5000568937
Encompass_Preferred
Pyrazinamide-d3 (CAS No 1432059-16-9) is a deuterium-labeled derivative used in biomedical research as a stable isotope for various studies Originating from antimicrobial research it is primarily utilized to investigate Mycobacterium tuberculosis the causative agent of tuberculosis Pyrazinamide-d3 acts by disrupting the bacterial membrane potential and interfering with fatty acid synthase I which is critical for the bacterium s survival and replication In vitro studies frequently demonstrate its activity in the nanomolar to low micromolar range making it effective for precise quantitative analysis This compound is commonly applied in cell models animal studies and drug screening assays to measure drug metabolism and pharmacokinetics Experimental concentrations typically depend on the specific research design allowing for flexibility across various experimental frameworks ultimately aiding in the development of novel therapeutics against bacterial infections
Catalog No. 50-005-68937 Supplier Apexbio Technology LLC Supplier No. SL10541
Pyrazinamide-d3 (CAS No 1432059-16-9) is a deuterium-labeled derivative used in biomedical research as a stable isotope for various studies Originating from antimicrobial research it is primarily utilized to investigate Mycobacterium tuberculosis the causative agent of tuberculosis Pyrazinamide-d3 acts by disrupting the bacterial membrane potential and interfering with fatty acid synthase I which is critical for the bacterium s survival and replication In vitro studies frequently demonstrate its activity in the nanomolar to low micromolar range making it effective for precise quantitative analysis This compound is commonly applied in cell models animal studies and drug screening assays to measure drug metabolism and pharmacokinetics Experimental concentrations typically depend on the specific research design allowing for flexibility across various experimental frameworks ultimately aiding in the development of novel therapeutics against bacterial infections