Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyrazinamide, MP Biomedicals™

Catalog No. MP219547380 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219547380 100 g
MP219547310 10 g
MP219547325 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP219547380 Supplier MP Biomedicals, Inc Supplier No. 0219547380
Add to Cart
Add to Cart

Pyrazinamide ≥99%, White Powder

  • It is used therapeutically as an antitubercular agent.
  • It is used to form polymeric copper complexes, create pyrazine carboxamide scaffolds useful as FXs inhibitors, and as a component of mycobacteria identification kits.
  • It is used to study liver toxicity prevention and mechanisms of resistance.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-96-4
Molecular Formula C5H5N3O
Molecular Weight (g/mol) 123.12
MDL Number MFCD00006132
InChI Key IPEHBUMCGVEMRF-UHFFFAOYSA-N

Specifications

Melting Point 185°C to 195°C
Color White
pH 5.6
Quantity 100 g
Beilstein 112306
Synonym Pyrazinecarboxamide, Pyrazinoic acid amide
Solubility Information Soluble in water (50mg/mL-clear).
Physical Form Powder
RTECSNumber UQ2275000
storageNote1 Store at Room Temperature (15-30°C).
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.