Learn More
Medchemexpress LLC Pyrazinamide (Standard) | 98-96-4 | MFCD00006132 | 99.99% | 250 MG
SDP
Catalog No. 5000406139
Encompass_Preferred
Pyrazinamide (Standard) is the analytical standard for Pyrazinamide, intended for research and analytical applications. This potent, orally active antitubercular antibiotic is a proagent, converted to pyrazinoic acid (POA) by PZase/nicotinamidase in *M. tuberculosis*. It serves as a reference standard for assay and is commonly utilized in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Analytical standard for Pyrazinamide
  • Intended for research and analytical applications
  • Potent and orally active antitubercular antibiotic
  • Proagent converted to active form pyrazinoic acid (POA)
  • Used in qualitative research experiments
  • Used in quantitative research experiments
  • Used in methodological research experiments
  • Commonly used in HPLC, GC, and MS
Catalog No. 50-004-06139 Supplier Medchemexpress LLC Supplier No. 250MGHYB0271R
Pyrazinamide (Standard) is the analytical standard for Pyrazinamide, intended for research and analytical applications. This potent, orally active antitubercular antibiotic is a proagent, converted to pyrazinoic acid (POA) by PZase/nicotinamidase in *M. tuberculosis*. It serves as a reference standard for assay and is commonly utilized in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Analytical standard for Pyrazinamide
  • Intended for research and analytical applications
  • Potent and orally active antitubercular antibiotic
  • Proagent converted to active form pyrazinoic acid (POA)
  • Used in qualitative research experiments
  • Used in quantitative research experiments
  • Used in methodological research experiments
  • Commonly used in HPLC, GC, and MS