Learn More
Medchemexpress LLC Pyrazinamine | 5049-61-6 | 99.86% | 95.10 | 100 G
SDP
Catalog No. 5000404455
Encompass_Preferred
Pyrazinamine is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is also a synthetic intermediate useful for pharmaceutical synthesis.
  • Suitable for life science research.
  • Can be used as a biological material or organic compound.
  • A synthetic intermediate for pharmaceutical synthesis.
  • Purity of 99.86%.
  • Appears as an off-white to light brown solid.
  • Specific storage conditions for solid and in solvent forms.
Catalog No. 50-004-04455 Supplier Medchemexpress LLC Supplier No. HY34068100G
Pyrazinamine is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is also a synthetic intermediate useful for pharmaceutical synthesis.
  • Suitable for life science research.
  • Can be used as a biological material or organic compound.
  • A synthetic intermediate for pharmaceutical synthesis.
  • Purity of 99.86%.
  • Appears as an off-white to light brown solid.
  • Specific storage conditions for solid and in solvent forms.