Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pyrazinamine | 5049-61-6 | 99.9% | 95.10 | 25 G
SDP

Catalog No. 5000403066
Encompass_Preferred

Pyrazinamine is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is also described as a synthetic intermediate useful for pharmaceutical synthesis. For research use only.

  • Off-white to light brown solid appearance
  • Molecular formula C4H5N3
  • Shipping at room temperature in continental US
  • Storage at 4°C, protected from light

Catalog No. 50-004-03066 Supplier Medchemexpress LLC Supplier No. 25GHY34068
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pyrazinamine is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is also described as a synthetic intermediate useful for pharmaceutical synthesis. For research use only.

  • Off-white to light brown solid appearance
  • Molecular formula C4H5N3
  • Shipping at room temperature in continental US
  • Storage at 4°C, protected from light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.