Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pyrazinamine | 5049-61-6 | 99.9% | 95.10 | 500 G
SDP

Supplier:  Medchemexpress LLC HY34068500G

Encompass_Preferred

Pyrazinamine is a biochemical reagent that can be used as a biological material or organic compound for life science related research. 2-Aminopyrazine is a synthetic intermediate useful for pharmaceutical synthesis.

  • Appearance: Solid
  • Color: Off-white to light brown
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: 4°C, protect from light; in solvent: -80°C, 6 months; -20°C, 1 month (protect from light)

Catalog No. 50-004-03620


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.