Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pyrazine-2,3-dicarboxylic acid | 89-01-0 | MFCD00006131 | 100.0% | 168.11 | C6H4N2O4 | 25 G
SDP

Catalog No. 5000302681
Encompass_Preferred

Pyrazine-2,3-dicarboxylic acid is an organic biochemical reagent used as a reference standard and building block in life-science research, analytical chemistry, and synthetic applications. It is supplied as a white to off-white solid with high purity and defined storage recommendations for long-term stability.

  • High purity suitable for analytical and research use.
  • White to off-white solid, easy to handle and weigh.
  • Well-defined chemical identity (C6H4N2O4, MW 168.11).
  • Multiple pack sizes for small-scale or bulk needs.
  • Storage conditions support long-term stability of powder and solutions.

Catalog No. 50-003-02681 Supplier Medchemexpress LLC Supplier No. 25GHYW017105
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pyrazine-2,3-dicarboxylic acid is an organic biochemical reagent used as a reference standard and building block in life-science research, analytical chemistry, and synthetic applications. It is supplied as a white to off-white solid with high purity and defined storage recommendations for long-term stability.

  • High purity suitable for analytical and research use.
  • White to off-white solid, easy to handle and weigh.
  • Well-defined chemical identity (C6H4N2O4, MW 168.11).
  • Multiple pack sizes for small-scale or bulk needs.
  • Storage conditions support long-term stability of powder and solutions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.