Learn More
Medchemexpress LLC Pyrazine-2,3-dicarboxylic acid | 89-01-0 | MFCD00006131 | 100.0% | 168.11 g/mol | C6H4N2O4 | 1 KG
SDP
Catalog No. 5000300337
Encompass_Preferred
Pyrazine-2,3-dicarboxylic acid (CAS 89-01-0) is an organic biochemical reagent supplied as a powder for research and analytical use. It functions as a building block or analytical standard in organic synthesis and life-science experiments, with stability and handling guidance documented in the product safety data sheet (SDS).
  • High purity (99.97%) for analytical and synthetic work.
  • Molecular weight 168.11 g/mol; formula C6H4N2O4.
  • Available in multiple package sizes; bulk quantities by inquiry.
  • Stable as powder when stored at recommended temperatures (-20°C to 4°C).
  • Safety data sheet available in multiple languages.
Catalog No. 50-003-00337 Supplier Medchemexpress LLC Supplier No. HYW0171051KG
Pyrazine-2,3-dicarboxylic acid (CAS 89-01-0) is an organic biochemical reagent supplied as a powder for research and analytical use. It functions as a building block or analytical standard in organic synthesis and life-science experiments, with stability and handling guidance documented in the product safety data sheet (SDS).
  • High purity (99.97%) for analytical and synthetic work.
  • Molecular weight 168.11 g/mol; formula C6H4N2O4.
  • Available in multiple package sizes; bulk quantities by inquiry.
  • Stable as powder when stored at recommended temperatures (-20°C to 4°C).
  • Safety data sheet available in multiple languages.