Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pyrazine-2,3-dicarboxylic acid | 89-01-0 | MFCD00006131 | 100.0% | 168.11 g/mol | C6H4N2O4 | 50 G
SDP

Catalog No. 5000299887
Encompass_Preferred

Pyrazine-2,3-dicarboxylic acid is a small-molecule biochemical reagent used in life-science research and analytical applications. It is supplied as a dry powder for use as an organic reagent or biological-material component in synthesis and analytical workflows.

  • High purity suitable for analytical and research use.
  • Molecular formula C6H4N2O4 and molecular weight 168.11 g/mol.
  • Soluble in DMSO at up to 100 mg/mL; ultrasonic assistance recommended; hygroscopic DMSO may affect solubility.
  • Powder storage: -20°C (3 years) or 4°C (2 years); solution storage: -80°C (6 months) or -20°C (1 month).
  • Available in multiple bulk package sizes for scalability.

Catalog No. 50-002-99887 Supplier Medchemexpress LLC Supplier No. HYW01710550G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pyrazine-2,3-dicarboxylic acid is a small-molecule biochemical reagent used in life-science research and analytical applications. It is supplied as a dry powder for use as an organic reagent or biological-material component in synthesis and analytical workflows.

  • High purity suitable for analytical and research use.
  • Molecular formula C6H4N2O4 and molecular weight 168.11 g/mol.
  • Soluble in DMSO at up to 100 mg/mL; ultrasonic assistance recommended; hygroscopic DMSO may affect solubility.
  • Powder storage: -20°C (3 years) or 4°C (2 years); solution storage: -80°C (6 months) or -20°C (1 month).
  • Available in multiple bulk package sizes for scalability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.