Learn More
Medchemexpress LLC Pyrazine-2,3-dicarboxylic acid | 89-01-0 | MFCD00006131 | 100.0% | 168.11 g/mol | C6H4N2O4 | 500 G
SDP
Catalog No. 5000300545
Encompass_Preferred
Pyrazine-2,3-dicarboxylic acid is a small organic compound and biochemical reagent (CAS 89-01-0) used in life-science research. It is supplied as a solid powder with high purity and is commonly used as a synthetic building block, analytical reference, and reagent in biochemical assays and organic synthesis.
  • High purity suitable for analytical and synthetic use.
  • Solid powder form for easy handling and storage.
  • Useful as a synthetic building block for pyrazine derivatives.
  • Suitable as an analytical reference standard.
  • Compatible with common organic synthesis and assay workflows.
  • Characterized molecular weight and formula for precise calculations.
Catalog No. 50-003-00545 Supplier Medchemexpress LLC Supplier No. HYW017105500G
Pyrazine-2,3-dicarboxylic acid is a small organic compound and biochemical reagent (CAS 89-01-0) used in life-science research. It is supplied as a solid powder with high purity and is commonly used as a synthetic building block, analytical reference, and reagent in biochemical assays and organic synthesis.
  • High purity suitable for analytical and synthetic use.
  • Solid powder form for easy handling and storage.
  • Useful as a synthetic building block for pyrazine derivatives.
  • Suitable as an analytical reference standard.
  • Compatible with common organic synthesis and assay workflows.
  • Characterized molecular weight and formula for precise calculations.