Learn More
Medchemexpress LLC Pyrazine-2-amidoxime | 51285-05-3 | MFCD02258397 | 99.9% | 138.13 g/mol | C5H6N4O | 250 MG
SDP
Catalog No. 5000303490
Encompass_Preferred
Pyrazine-2-amidoxime is a small-molecule amidoxime used as a research reagent and synthetic intermediate. It is a structural analogue of pyrazine-2-carboxamide with reported antimicrobial activity against Candida albicans, fungal pathogens, and Gram-positive bacteria. The compound is a white to off-white solid with formula C5H6N4O and molecular weight 138.13 g/mol.
  • High stated purity (99.9%).
  • Available in multiple small-scale sizes suitable for laboratory use.
  • Water-soluble with warming and ultrasonic assistance.
  • Store sealed and protect from moisture for best stability.
  • For research use only; filter-sterilize aqueous stocks before use.
Catalog No. 50-003-03490 Supplier Medchemexpress LLC Supplier No. 250MGHYW031471A
Pyrazine-2-amidoxime is a small-molecule amidoxime used as a research reagent and synthetic intermediate. It is a structural analogue of pyrazine-2-carboxamide with reported antimicrobial activity against Candida albicans, fungal pathogens, and Gram-positive bacteria. The compound is a white to off-white solid with formula C5H6N4O and molecular weight 138.13 g/mol.
  • High stated purity (99.9%).
  • Available in multiple small-scale sizes suitable for laboratory use.
  • Water-soluble with warming and ultrasonic assistance.
  • Store sealed and protect from moisture for best stability.
  • For research use only; filter-sterilize aqueous stocks before use.