Learn More
Medchemexpress LLC Pyrazine-2-amidoxime | 51285-05-3 | MFCD02258397 | 99.9% | 138.13 g/mol | C5H6N4O | 5 G
SDP
Catalog No. 5000299341
Encompass_Preferred
Pyrazine-2-amidoxime is a small-molecule research chemical and a structural analogue of pyrazine-2-carboxamide with reported antimicrobial activity against Candida albicans and Gram-positive bacteria. It is used as a synthetic intermediate in pharmaceutical research and is supplied for research use only.
  • Reported antimicrobial activity against Candida albicans and Gram-positive bacteria.
  • Useful synthetic intermediate for pharmaceutical research.
  • High purity (99.9%) suitable for analytical and synthetic work.
  • Molecular formula C5H6N4O; molecular weight 138.13 g/mol.
  • Supplied in multiple pack sizes, including a 5 g option.
  • For research use only; not for human or veterinary use.
Catalog No. 50-002-99341 Supplier Medchemexpress LLC Supplier No. HYW031471A5G
Pyrazine-2-amidoxime is a small-molecule research chemical and a structural analogue of pyrazine-2-carboxamide with reported antimicrobial activity against Candida albicans and Gram-positive bacteria. It is used as a synthetic intermediate in pharmaceutical research and is supplied for research use only.
  • Reported antimicrobial activity against Candida albicans and Gram-positive bacteria.
  • Useful synthetic intermediate for pharmaceutical research.
  • High purity (99.9%) suitable for analytical and synthetic work.
  • Molecular formula C5H6N4O; molecular weight 138.13 g/mol.
  • Supplied in multiple pack sizes, including a 5 g option.
  • For research use only; not for human or veterinary use.