Learn More
Medchemexpress LLC Pyrazine | 290-37-9 | 100.0% | 80.09 | 1 KG
SDP
Catalog No. 5000454140
Encompass_Preferred
Pyrazine is an aromatic nitrogen-containing heterocyclic compound with two nitrogen atoms, known for its electron-deficient properties. It acts as a crucial pharmacophore in various drugs and forms the core skeleton of derivatives with anticancer, antimycobacterial, antithrombotic, and anti-inflammatory activities.
  • Undergoes Chichibabin reaction with sodamide to produce 2-aminopyrazine.
  • Reacts with H2O2/CH3COOH to yield mono and dioxide products.
  • Can be synthesized through catalytic dehydrogenation of ethanolamine.
  • Forms the core skeleton of derivatives with anticancer, antimycobacterial, antithrombotic, and anti-inflammatory activities.
Catalog No. 50-004-54140 Supplier Medchemexpress LLC Supplier No. HYW0130401KG
Pyrazine is an aromatic nitrogen-containing heterocyclic compound with two nitrogen atoms, known for its electron-deficient properties. It acts as a crucial pharmacophore in various drugs and forms the core skeleton of derivatives with anticancer, antimycobacterial, antithrombotic, and anti-inflammatory activities.
  • Undergoes Chichibabin reaction with sodamide to produce 2-aminopyrazine.
  • Reacts with H2O2/CH3COOH to yield mono and dioxide products.
  • Can be synthesized through catalytic dehydrogenation of ethanolamine.
  • Forms the core skeleton of derivatives with anticancer, antimycobacterial, antithrombotic, and anti-inflammatory activities.