Learn More
Medchemexpress LLC Pyrazine | 290-37-9 | 99.97% | 80.09 | 100 G
SDP
Catalog No. 5000451199
Encompass_Preferred
Pyrazine is an aromatic nitrogen-containing heterocyclic compound with two nitrogen atoms, known for its electron-deficient property. It serves as a key pharmacophore in various drugs and acts as the core skeleton of derivatives with anticancer, antimycobacterial, antithrombotic, and anti-inflammatory activities.
  • Key pharmacophore in various drugs
  • Core skeleton for derivatives with anticancer, antimycobacterial, antithrombotic, and anti-inflammatory activities
  • Undergoes Chichibabin reaction to form 2-aminopyrazine
  • Reacts with H2O2/CH3COOH to form mono and dioxide products
  • Can be synthesized via catalytic dehydrogenation of ethanolamine
  • Monobasic compound with a pKa of 0.65
Catalog No. 50-004-51199 Supplier Medchemexpress LLC Supplier No. HYW013040100G
Pyrazine is an aromatic nitrogen-containing heterocyclic compound with two nitrogen atoms, known for its electron-deficient property. It serves as a key pharmacophore in various drugs and acts as the core skeleton of derivatives with anticancer, antimycobacterial, antithrombotic, and anti-inflammatory activities.
  • Key pharmacophore in various drugs
  • Core skeleton for derivatives with anticancer, antimycobacterial, antithrombotic, and anti-inflammatory activities
  • Undergoes Chichibabin reaction to form 2-aminopyrazine
  • Reacts with H2O2/CH3COOH to form mono and dioxide products
  • Can be synthesized via catalytic dehydrogenation of ethanolamine
  • Monobasic compound with a pKa of 0.65