Learn More
Medchemexpress LLC Pyrazine | 290-37-9 | 99.97% | 80.09 | 500 G
SDP
Supplier:  Medchemexpress LLC HYW013040500G
Encompass_Preferred
Pyrazine is an aromatic nitrogen-containing heterocyclic compound with two nitrogen atoms, known for its electron-deficient property. It serves as a key pharmacophore in various drugs and acts as the core skeleton of derivatives with anticancer, antimycobacterial, antithrombotic, and anti-inflammatory activities.
  • Undergoes Chichibabin reaction with sodamide to form 2-aminopyrazine.
  • Reacts with 30% H2O2/CH3COOH at 70-80 °C to form mono and dioxide products.
  • Can be synthesized via catalytic dehydrogenation of ethanolamine or via double condensation of a 1,2-dicarbonyl compound with equimolar 1,2-diamine followed by oxidation.
  • It is a monobasic compound with a pKa of 0.65, protonated at N-1, and only undergoes N,N-diprotonation in very strong acidic media.
Catalog No. 50-004-52000