Learn More
Medchemexpress LLC Pyrazine-d4 | 1758-62-9 | 84.11 | 1 MG
SDP
Catalog No. 5000454066
Encompass_Preferred
Pyrazine-d4 is a deuterium-labeled pyrazine designed for research applications. This compound is primarily used as a tracer for quantitation during drug development processes. The incorporation of stable heavy isotopes, such as deuterium, into drug molecules has been shown to affect pharmacokinetic and metabolic profiles.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-54066 Supplier Medchemexpress LLC Supplier No. HYW013040S1MG
Pyrazine-d4 is a deuterium-labeled pyrazine designed for research applications. This compound is primarily used as a tracer for quantitation during drug development processes. The incorporation of stable heavy isotopes, such as deuterium, into drug molecules has been shown to affect pharmacokinetic and metabolic profiles.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS