Learn More
Medchemexpress LLC Pyrazino[1,2-a]benzimidazol-1(2H)-one, 3,4-dihydro-2-[4-[4-(trifluoromethoxy)phenoxy]butyl]- | 2671063-84-4 | 98.0% | C21H20F3N3O3 | 10MG
SDP
Catalog No. 5000260935
Encompass_Preferred
mGluR2 modulator 1 is a research-grade positive allosteric modulator of the metabotropic glutamate receptor 2 (mGluR2), used in in vitro and in vivo pharmacology studies. It is reported to be brain-penetrant and displays potent receptor activity suitable for mechanistic and preclinical research.
  • Potent mGluR2 positive allosteric modulator (EC50 ≈ 0.03 μM).
  • Brain-penetrant profile suitable for in vivo studies.
  • High purity for research applications.
  • Soluble in DMSO (50 mg/mL); in vivo vehicle options documented.
  • Stable when stored protected from light at recommended temperatures.
  • Supplied as a white to off-white solid for handling and formulation.
Catalog No. 50-002-60935 Supplier Medchemexpress LLC Supplier No. HY13063010MG
mGluR2 modulator 1 is a research-grade positive allosteric modulator of the metabotropic glutamate receptor 2 (mGluR2), used in in vitro and in vivo pharmacology studies. It is reported to be brain-penetrant and displays potent receptor activity suitable for mechanistic and preclinical research.
  • Potent mGluR2 positive allosteric modulator (EC50 ≈ 0.03 μM).
  • Brain-penetrant profile suitable for in vivo studies.
  • High purity for research applications.
  • Soluble in DMSO (50 mg/mL); in vivo vehicle options documented.
  • Stable when stored protected from light at recommended temperatures.
  • Supplied as a white to off-white solid for handling and formulation.