Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pyrazino[2,1-a]pyrido[2,3-c]benzazepine, 1,2,3,4,10,14b-hexahydro-2-methyl-, (14bS)- | 61337-87-9 | 99.8% | 25 MG
SDP

Catalog No. 5000450582
Encompass_Preferred

(S)-Mirtazapine ((S)-Org3770) is an S(+)-enantiomer of Mirtazapine. It exhibits pronociceptive properties in an animal model of acute thermal nociception and acts as a stereoselective 5-HT2 receptor antagonist. It is metabolized by CYP2D6 and CYP1A2. The product is intended for research use only and is not sold to patients.

  • S(+)-enantiomer of mirtazapine
  • Exhibits pronociceptive properties in an animal model
  • Acts as a stereoselective 5-HT2 receptor antagonist
  • Metabolized by CYP2D6 and CYP1A2
  • Purity: 99.8%
  • Enantiomeric excess: 98.35%
  • Molecular weight: 265.35
  • Molecular formula: C17H19N3
  • Appearance: Solid, off-white to light yellow
  • Solubility in DMSO: ≥ 100 mg/mL
  • Intended for research use only

Catalog No. 50-004-50582 Supplier Medchemexpress LLC Supplier No. 25MGHYB0352A
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(S)-Mirtazapine ((S)-Org3770) is an S(+)-enantiomer of Mirtazapine. It exhibits pronociceptive properties in an animal model of acute thermal nociception and acts as a stereoselective 5-HT2 receptor antagonist. It is metabolized by CYP2D6 and CYP1A2. The product is intended for research use only and is not sold to patients.

  • S(+)-enantiomer of mirtazapine
  • Exhibits pronociceptive properties in an animal model
  • Acts as a stereoselective 5-HT2 receptor antagonist
  • Metabolized by CYP2D6 and CYP1A2
  • Purity: 99.8%
  • Enantiomeric excess: 98.35%
  • Molecular weight: 265.35
  • Molecular formula: C17H19N3
  • Appearance: Solid, off-white to light yellow
  • Solubility in DMSO: ≥ 100 mg/mL
  • Intended for research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.