Learn More
Medchemexpress LLC Pyrazole-3-carboxylic acid | 1621-91-6 | MFCD00077436 | 112.09 | 100 G
SDP
Catalog No. 5000445883
Encompass_Preferred
Pyrazole-3-carboxylic acid is an alternative anchoring group to carboxylic acid in phthalocyanine dyes, exhibiting flexible coordination modes. It can chelate to metal ions to form stable complexes via its N and O atoms. This compound can be utilized in the synthesis of derivatives with various biological activities.
  • Serves as an alternative anchoring group in phthalocyanine dyes
  • Exhibits flexible coordination modes with metal ions
  • Forms stable complexes with metal ions via N and O atoms
  • Used in the synthesis of derivatives with antibacterial activity
  • Used in the synthesis of derivatives with anti-inflammatory activity
  • Used in the synthesis of derivatives with ulcerogenic liability
  • Used in the synthesis of derivatives with antiproliferative activities
Catalog No. 50-004-45883 Supplier Medchemexpress LLC Supplier No. HYW002328100G
Add to Cart
Add to Cart
Pyrazole-3-carboxylic acid is an alternative anchoring group to carboxylic acid in phthalocyanine dyes, exhibiting flexible coordination modes. It can chelate to metal ions to form stable complexes via its N and O atoms. This compound can be utilized in the synthesis of derivatives with various biological activities.
  • Serves as an alternative anchoring group in phthalocyanine dyes
  • Exhibits flexible coordination modes with metal ions
  • Forms stable complexes with metal ions via N and O atoms
  • Used in the synthesis of derivatives with antibacterial activity
  • Used in the synthesis of derivatives with anti-inflammatory activity
  • Used in the synthesis of derivatives with ulcerogenic liability
  • Used in the synthesis of derivatives with antiproliferative activities