Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyrazole 98.0+%, TCI America™
SDP

Catalog No. P054625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P054625G 25 g
P0546250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P054625G Supplier TCI America Supplier No. P054625G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 288-13-1
Molecular Formula C3H4N2
Molecular Weight (g/mol) 68.079
MDL Number MFCD00005234
InChI Key WTKZEGDFNFYCGP-UHFFFAOYSA-N
Synonym pyrazole, 1,2-diazole, 1h-pyrazol, pyrazol, unii-3qd5kjz7zj, 3qd5kjz7zj, diazole, hpz, 1-h-pyrazole, pyrazol#1
PubChem CID 1048
ChEBI CHEBI:17241
IUPAC Name 1H-pyrazole
SMILES C1=CNN=C1

Specifications

Melting Point 69°C
Color White-Yellow
Boiling Point 188°C
Quantity 25 g
Formula Weight 68.08
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Pyrazole
EINECSNumber (5)-5624
RTECSNumber UQ4900000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.