Learn More
Medchemexpress LLC Pyrazoline-based chloroacetamide | 99.6% | 404.89 g·mol⁻¹ | C24H21ClN2O2 | 10 MG
SDP
Catalog No. 5000288793
Encompass_Preferred
MM-02-57 is a covalent small-molecule recruiter that targets the DDB1 adaptor to engage the CUL4A ubiquitin ligase complex for targeted protein degradation studies. It is provided as a solid powder intended for laboratory research use only.
  • Covalent recruiter of DDB1 for CUL4A adaptor engagement.
  • Designed for use in targeted protein degradation and chemoproteomics research.
  • Pyrazoline-based chloroacetamide scaffold with cysteine-reactive electrophile.
  • Molecular weight 404.89 g·mol⁻¹; chemical formula C24H21ClN2O2.
  • Supplied as a solid powder with cold storage recommendations for stability.
  • High reported purity from distributor analyses, suitable for biochemical assays.
Catalog No. 50-002-88793 Supplier Medchemexpress LLC Supplier No. HY16116610MG
MM-02-57 is a covalent small-molecule recruiter that targets the DDB1 adaptor to engage the CUL4A ubiquitin ligase complex for targeted protein degradation studies. It is provided as a solid powder intended for laboratory research use only.
  • Covalent recruiter of DDB1 for CUL4A adaptor engagement.
  • Designed for use in targeted protein degradation and chemoproteomics research.
  • Pyrazoline-based chloroacetamide scaffold with cysteine-reactive electrophile.
  • Molecular weight 404.89 g·mol⁻¹; chemical formula C24H21ClN2O2.
  • Supplied as a solid powder with cold storage recommendations for stability.
  • High reported purity from distributor analyses, suitable for biochemical assays.