Learn More
Medchemexpress LLC Pyrazolo[1,5-a]pyrimidine-2,6-diamine, N2,N6,N6,5,7-pentamethyl-3-(phenylsulfonyl)- | 1220646-23-0 | 99.3% | 359.45 g/mol | C17H21N5O2S | 100 MG
SDP
Catalog No. 5000248500
Encompass_Preferred
AVN-492 is a potent, highly selective antagonist of the serotonin 5-HT6 receptor used as a research tool in CNS pharmacology and cognitive/neurodegenerative studies. It exhibits picomolar affinity for 5-HT6 and is supplied with high analytical purity for in vitro and in vivo research.
  • High potency, binds 5-HT6 receptor with Ki = 91 pM.
  • Strong selectivity, low affinity at 5-HT2B (Ki = 170 nM).
  • High purity suitable for research (LCMS purity 99.3%).
  • Available in multiple package sizes, including a 100 mg option.
  • Supplied as solid or DMSO solution for flexible experimental use.
Catalog No. 50-002-48500 Supplier Medchemexpress LLC Supplier No. HY101924100MG
AVN-492 is a potent, highly selective antagonist of the serotonin 5-HT6 receptor used as a research tool in CNS pharmacology and cognitive/neurodegenerative studies. It exhibits picomolar affinity for 5-HT6 and is supplied with high analytical purity for in vitro and in vivo research.
  • High potency, binds 5-HT6 receptor with Ki = 91 pM.
  • Strong selectivity, low affinity at 5-HT2B (Ki = 170 nM).
  • High purity suitable for research (LCMS purity 99.3%).
  • Available in multiple package sizes, including a 100 mg option.
  • Supplied as solid or DMSO solution for flexible experimental use.