Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyrazolo[1,5-a]pyrimidine-2-carboxylic acid, 97%, Thermo Scientific™

Catalog No. AC434340010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC434340010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC434340010 Supplier Thermo Scientific Chemicals Supplier No. 434340010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 378211-85-9
Molecular Formula C7H5N3O2
Molecular Weight (g/mol) 163.14
MDL Number MFCD00297221
InChI Key DZPAOAZDQHZRGG-UHFFFAOYSA-N
Synonym pyrazolo 1,5-a pyrimidine-2-carboxylic acid, 8-hydropyrazolo 1,5-a pyrimidine-2-carboxylic acid, 2-pyrazolo 1,5-a pyrimidinecarboxylic acid, pyrazolo 1,5-a pyrimidine-2-carboxylicacid, acmc-1afu0, pyrazolo 1,5-a pyrimidinecarboxylic acid, pyrazolo 1?5-a pyrimidine-2-carboxylic acid, pyrazolo 1,5-a pyrimidine-2-carboxylic acid 9ci, 2-methyl-3-2-trimethylsilyl ethoxy methyl-3h-imidazo 4,5-b pyridin-6-yl boronic acid
PubChem CID 670824
IUPAC Name pyrazolo[1,5-a]pyrimidine-2-carboxylic acid
SMILES C1=CN2C(=CC(=N2)C(=O)O)N=C1

Specifications

Melting Point 289°C to 291°C
Color Orange to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 163.14
Percent Purity 97%
Physical Form Crystalline Powder or Powder
Chemical Name or Material Pyrazolo[1, 5-a]pyrimidine-2-carboxylic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.