Learn More
Medchemexpress LLC Pyrazolo[1,5-a]pyrimidine-3-carbonitrile, 4,7-dihydro-6-isopropyl-7-oxo-5-phenyl hydrochloride | 2095432-28-1 | 100.0% | 314.77 g/mol | C16H15ClN4O | 1 MG
SDP
Catalog No. 5000213936
Encompass_Preferred
CPI-455 hydrochloride is the hydrochloride salt of a potent pan-KDM5 histone demethylase inhibitor with reported low-nanomolar activity against KDM5A. It is intended for research use in epigenetics and cancer biology to elevate H3K4me3 levels and probe mechanisms of drug-tolerant persister cells. Supplied as a high-purity reagent for in vitro studies.
  • Potent pan-KDM5 inhibition with low-nanomolar activity against KDM5A.
  • Elevates global H3K4me3 levels in cellular assays.
  • High purity suitable for research-grade experiments.
  • Available in small milligram pack sizes for screening and assays.
  • Hydrochloride salt form improves aqueous solubility for formulation.
Catalog No. 50-002-13936 Supplier Medchemexpress LLC Supplier No. HY100421A1MG
CPI-455 hydrochloride is the hydrochloride salt of a potent pan-KDM5 histone demethylase inhibitor with reported low-nanomolar activity against KDM5A. It is intended for research use in epigenetics and cancer biology to elevate H3K4me3 levels and probe mechanisms of drug-tolerant persister cells. Supplied as a high-purity reagent for in vitro studies.
  • Potent pan-KDM5 inhibition with low-nanomolar activity against KDM5A.
  • Elevates global H3K4me3 levels in cellular assays.
  • High purity suitable for research-grade experiments.
  • Available in small milligram pack sizes for screening and assays.
  • Hydrochloride salt form improves aqueous solubility for formulation.