Learn More
Medchemexpress LLC Pyrazolo[1,5-a]pyrimidine-3-carboxamide derivative | 1831144-46-7 | 99.9% | 612.03 g/mol | C28H28ClF2N9O3 | 5MG
SDP
Catalog No. 5000235201
Encompass_Preferred
iJak-381 is a small-molecule Janus kinase (JAK) inhibitor provided for research use only. It is supplied as a solid powder and is used in preclinical studies to investigate JAK-mediated signaling and pulmonary inflammation. Supplier documentation lists chemical, purity, and storage information.
Catalog No. 50-002-35201 Supplier Medchemexpress LLC Supplier No. HY1250195MG
iJak-381 is a small-molecule Janus kinase (JAK) inhibitor provided for research use only. It is supplied as a solid powder and is used in preclinical studies to investigate JAK-mediated signaling and pulmonary inflammation. Supplier documentation lists chemical, purity, and storage information.