Learn More
Medchemexpress LLC Pyrazoloadenine | 2380-63-4 | 100.0% | 135.13 | 25 G
SDP
Catalog No. 5000402378
Encompass_Preferred
Pyrazoloadenine is a potent inhibitor of the RET (REarranged during Transfection) lung cancer oncoprotein, demonstrating anticancer activity. It is intended for research use only and not for sale to patients.
  • Potent RET lung cancer oncoprotein inhibitor.
  • Shows anticancer activity.
  • Inhibits a wide range of protein kinases, including BTK, CDK1/2, Src kinase, and RIPK1.
Catalog No. 50-004-02378 Supplier Medchemexpress LLC Supplier No. 25GHY34595
Pyrazoloadenine is a potent inhibitor of the RET (REarranged during Transfection) lung cancer oncoprotein, demonstrating anticancer activity. It is intended for research use only and not for sale to patients.
  • Potent RET lung cancer oncoprotein inhibitor.
  • Shows anticancer activity.
  • Inhibits a wide range of protein kinases, including BTK, CDK1/2, Src kinase, and RIPK1.