Learn More
Associates of Cape Cod Inc LAL Reagent Water, 5.5 mL, 10 bottles per pack, Endotoxin Level < 0.001 EU/mL, Glucan Level < 1.56 pg/mL, not for human or animal injection
SDP
Catalog No. NC2056375
Encompass
This LAL Reagent Water (LRW) from Associates of Cape Cod, Inc. is specially processed for critical applications in Bacterial Endotoxin Testing (BET). It serves as an ideal solution for reconstituting LAL reagents and Control Standard Endotoxin (CSE), as well as for diluting samples and standards in LAL assays. Its exceptional low levels of endotoxin and (1→3)-β-D-glucan minimize contamination risks, supporting compliance and reducing Out-of-Specification (OOS) investigations.
  • Product type: LAL Reagent Water
  • Endotoxin level: Less than 0.001 EU/mL
  • Glucan level: Less than 1.56 pg/mL
  • Volume per bottle: 5.5 mL
  • Bottles per pack: 10
  • Primary application: Reconstitution of LAL reagents and CSE
  • Secondary application: Dilution of samples and standards for LAL assays
  • Usage restriction: Not for human or animal injection
Catalog No. NC2056375 Supplier Associates of Cape Cod Inc Supplier No. W005110
This LAL Reagent Water (LRW) from Associates of Cape Cod, Inc. is specially processed for critical applications in Bacterial Endotoxin Testing (BET). It serves as an ideal solution for reconstituting LAL reagents and Control Standard Endotoxin (CSE), as well as for diluting samples and standards in LAL assays. Its exceptional low levels of endotoxin and (1→3)-β-D-glucan minimize contamination risks, supporting compliance and reducing Out-of-Specification (OOS) investigations.
  • Product type: LAL Reagent Water
  • Endotoxin level: Less than 0.001 EU/mL
  • Glucan level: Less than 1.56 pg/mL
  • Volume per bottle: 5.5 mL
  • Bottles per pack: 10
  • Primary application: Reconstitution of LAL reagents and CSE
  • Secondary application: Dilution of samples and standards for LAL assays
  • Usage restriction: Not for human or animal injection