Learn More
Medchemexpress LLC Pyrene-1,3,6,8-tetrasulfonic acid (tetrasodium) | 59572-10-0 | 99.0% | 50 G
SDP
Catalog No. 5000410517
Encompass_Preferred
Pyrene-1,3,6,8-tetrasulfonic acid tetrasodium is a fluorescent dye and pH indicator. It functions as a ligand of multifunctional metal-organic frameworks and has been used to detect CO2 release.
  • Fluorescent dye
  • pH indicator
  • Ligand of multifunctional metal-organic framework
  • Used to detect CO2 release
Catalog No. 50-004-10517 Supplier Medchemexpress LLC Supplier No. HYD014250G
Pyrene-1,3,6,8-tetrasulfonic acid tetrasodium is a fluorescent dye and pH indicator. It functions as a ligand of multifunctional metal-organic frameworks and has been used to detect CO2 release.
  • Fluorescent dye
  • pH indicator
  • Ligand of multifunctional metal-organic framework
  • Used to detect CO2 release