Learn More
Medchemexpress LLC Pyrene | 129-00-0 | MFCD00004136 | 99.7% | 202.25 g/mol | C16H10 | 25 G
SDP
Catalog No. 5000247640
Encompass_Preferred
Pyrene is a polycyclic aromatic hydrocarbon (PAH) commonly used as a fluorescent probe and biochemical reagent in research and analytical applications. It has molecular formula C16H10 and a molecular weight of 202.25 g/mol. Pyrene's strong fluorescence and well-characterized photophysical properties make it useful for studying molecular interactions, membrane dynamics, and environmental PAH behavior. Handle according to the safety data sheet.
Catalog No. 50-002-47640 Supplier Medchemexpress LLC Supplier No. HY10360925G
Pyrene is a polycyclic aromatic hydrocarbon (PAH) commonly used as a fluorescent probe and biochemical reagent in research and analytical applications. It has molecular formula C16H10 and a molecular weight of 202.25 g/mol. Pyrene's strong fluorescence and well-characterized photophysical properties make it useful for studying molecular interactions, membrane dynamics, and environmental PAH behavior. Handle according to the safety data sheet.