Learn More
Medchemexpress LLC Pyrene | 129-00-0 | MFCD00004136 | 99.8% | 202.25 g/mol | C16H10 | 10 G
SDP
Catalog No. 5000245154
Encompass_Preferred
Pyrene (C16H10) is a polycyclic aromatic hydrocarbon composed of four fused benzene rings. It is used as a fluorescent probe and as a model polycyclic aromatic hydrocarbon (PAH) for environmental, analytical, and materials research. Pyrene exhibits strong fluorescence in the blue region and is applied to study molecular interactions, aggregation, and surface phenomena. Handle with appropriate safety precautions because of potential health hazards associated with long-term exposure.
  • Polycyclic aromatic hydrocarbon with strong fluorescence.
  • Useful as a fluorescent probe for studying molecular interactions and microenvironments.
  • Common model compound for environmental and analytical studies of PAHs.
  • Solid reagent suitable for synthesis and analytical standard preparation.
  • Requires appropriate handling and personal protective equipment due to potential carcinogenic and mutagenic risks.
Catalog No. 50-002-45154 Supplier Medchemexpress LLC Supplier No. HY10360910G
Pyrene (C16H10) is a polycyclic aromatic hydrocarbon composed of four fused benzene rings. It is used as a fluorescent probe and as a model polycyclic aromatic hydrocarbon (PAH) for environmental, analytical, and materials research. Pyrene exhibits strong fluorescence in the blue region and is applied to study molecular interactions, aggregation, and surface phenomena. Handle with appropriate safety precautions because of potential health hazards associated with long-term exposure.
  • Polycyclic aromatic hydrocarbon with strong fluorescence.
  • Useful as a fluorescent probe for studying molecular interactions and microenvironments.
  • Common model compound for environmental and analytical studies of PAHs.
  • Solid reagent suitable for synthesis and analytical standard preparation.
  • Requires appropriate handling and personal protective equipment due to potential carcinogenic and mutagenic risks.