Learn More
Apexbio Technology LLC Pyrene azide 3, 25mg. Cas: MFCD: NA. Reagent for fluorescent labeling
SDP
Supplier:  Apexbio Technology LLC A812325
Encompass_Preferred
This pyrene azide is a derivative of pyrenebutyric acid. Contains triethylene glycol linker to facilitate dissolution during labeling reaction. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5030