Learn More
Medchemexpress LLC Pyrene-d10 | 1718-52-1 | C16D10 | 100 MG
SDP
Catalog No. 5000375099
Encompass_Preferred
Pyrene-d10 is the deuterium labeled Pyrene. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-75099 Supplier Medchemexpress LLC Supplier No. HY150627S100MG
Pyrene-d10 is the deuterium labeled Pyrene. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.