Learn More
Medchemexpress LLC Pyrene-d10 | 1718-52-1 | C16D10 | 50 MG
SDP
Catalog No. 5000375879
Encompass_Preferred
Pyrene-d10 is a deuterium labeled form of Pyrene, designed for research use only.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • For research applications only
Catalog No. 50-003-75879 Supplier Medchemexpress LLC Supplier No. HY150627S50MG
Pyrene-d10 is a deuterium labeled form of Pyrene, designed for research use only.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • For research applications only