Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyrene-d10 Solution, 500μg/mL in acetone, Ultra Scientific

Catalog No. USIST3701
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-IST-3701 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-IST-3701 Supplier Agilent Technologies Supplier No. IST3701
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chemical Identifiers

CAS 135-98-8 , 67-64-1
Molecular Formula C10H14
Molecular Weight (g/mol) 134.222
InChI Key ZJMWRROPUADPEA-UHFFFAOYSA-N
PubChem CID 8680
ChEBI CHEBI:35097
IUPAC Name butan-2-ylbenzene
SMILES CCC(C)C1=CC=CC=C1

Specifications

Physical Form Liquid
Color Colorless
Quantity 1 x 1 mL
Chemical Name or Material Pyrene-d10 Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.