Learn More
Medchemexpress LLC Pyrene maleimide | 1869968-64-8 | 98.1% | 410.46 g/mol | C26H22N2O3 | 10 MG
SDP
Catalog No. 5000297525
Encompass_Preferred
Pyrene maleimide is a polyaromatic fluorescent hydrocarbon bearing a terminal maleimide group that reacts covalently with thiol groups. It is provided as a solid for research and analytical applications, and includes solubility and storage recommendations to support use in labeling and fluorescence assays.
  • Fluorescent label for thiol-containing molecules.
  • Terminal maleimide group enables covalent thiol conjugation.
  • High purity (≈98.1%) suitable for analytical applications.
  • Soluble in DMSO: 100 mg/mL; may require ultrasonic agitation.
  • Stable at room temperature for extended storage; follow low-temperature storage for solutions.
  • Accompanied by analytical documentation, including COA, SDS, and spectral data.
Catalog No. 50-002-97525 Supplier Medchemexpress LLC Supplier No. HYD136410MG
Pyrene maleimide is a polyaromatic fluorescent hydrocarbon bearing a terminal maleimide group that reacts covalently with thiol groups. It is provided as a solid for research and analytical applications, and includes solubility and storage recommendations to support use in labeling and fluorescence assays.
  • Fluorescent label for thiol-containing molecules.
  • Terminal maleimide group enables covalent thiol conjugation.
  • High purity (≈98.1%) suitable for analytical applications.
  • Soluble in DMSO: 100 mg/mL; may require ultrasonic agitation.
  • Stable at room temperature for extended storage; follow low-temperature storage for solutions.
  • Accompanied by analytical documentation, including COA, SDS, and spectral data.