Learn More
Abcam Pyrene 1-sulfonyl chloride, 25MG
Catalog No. 50996113
Encompass_Preferred
MW 300.8 Da, Purity >85%. Amine reactive reagent for labeling peptides, proteins and other biological molecules. Fluorescence emission is environment-dependent.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-996-113 Supplier Abcam Supplier No. AB27505625MG
MW 300.8 Da, Purity >85%. Amine reactive reagent for labeling peptides, proteins and other biological molecules. Fluorescence emission is environment-dependent.
The product is subject to the following: Abcam Restricted Use Statement