Learn More
Abcam 1-Pyrenedecanoic acid, 10MG
Catalog No. 50996100
Encompass_Preferred
MW 372.5 Da, Purity >99%. Fluorescent lipid analog. Incorporates into membranes and lipid bilayers. Can be formed into micelles with other lipids (triglycerides, phospholipids). Used for fluorescent labeling, measurement of membrabe fluidity and lipase enzyme assays.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-996-100 Supplier Abcam Supplier No. AB27430910MG
MW 372.5 Da, Purity >99%. Fluorescent lipid analog. Incorporates into membranes and lipid bilayers. Can be formed into micelles with other lipids (triglycerides, phospholipids). Used for fluorescent labeling, measurement of membrabe fluidity and lipase enzyme assays.
The product is subject to the following: Abcam Restricted Use Statement