Learn More
AdipoGen Pyrenetetrasulfonic acid 4Na
SDP
Catalog No. 502560276
Encompass_Preferred
Chemical. CAS 59572-10-0. Formula C16H6Na4O12S4 . xH2O. MW 610.43 anhydrous basis. Synthetic Fluorescent probe and pH indicator. It fluoresces blue in acidic solutions and in acidic organelles but fluoresces green in more basic organelles. This compound is potentially useful as a coloring agent, biological stain or optical detecting reagent. It is used in water-in-oil-in-water emulsions and as a substrate for lipase activity. Spectral data lambdaex 375nm, lambdaem 404nm in water.
Catalog No. 50-256-0276 Supplier AdipoGen Supplier No. CDXP0021G010
Chemical. CAS 59572-10-0. Formula C16H6Na4O12S4 . xH2O. MW 610.43 anhydrous basis. Synthetic Fluorescent probe and pH indicator. It fluoresces blue in acidic solutions and in acidic organelles but fluoresces green in more basic organelles. This compound is potentially useful as a coloring agent, biological stain or optical detecting reagent. It is used in water-in-oil-in-water emulsions and as a substrate for lipase activity. Spectral data lambdaex 375nm, lambdaem 404nm in water.