Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ PYREX™ 24/29 Standard Taper Desiccator Stopcock with Hose Connector

Catalog No. 08624426 Shop All Corning Products
Change view
Click to view available options
Quantity:
1/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
08-624-426 1/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 08-624-426 Supplier Corning™ Supplier No. 3121STPK
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Replacement 24/29 standard taper desiccator stopcock with a tooled #10 hose connection

This is a replacement 24/29 Standard Taper stopcock with a tooled #10 hose connection. The plug material is PTFE, and is a suitable replacement for Cat. No. 3121-150, 3121-200 and 3121-250.

TRUSTED_SUSTAINABILITY

Specifications

Height (Metric) 11.68 cm
Height (English) 4.6 in.
Includes Hose connector
Product Type Desiccator Stopcock
Quantity 1/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.