Learn More
Corning™ PYREX™ Bleed Tubes With Capillary Tip
GREENER_CHOICE
Catalog No. C781519 Shop All Corning Products
Change view
Click to view available options
Material:
Borosilicate Glass
Catalog No. Material
C7815-19 Borosilicate Glass
1 options
Catalog No. C7815-19 Supplier Corning™ Supplier No. 781519
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
For bleeding gases, steam or liquids
These 7x280mm PYREX™ capillary tip bleed tubes are for bleeding gases, steam or liquids below the liquid surface of sidearm distillation flasks. This tube is also a replacement part for organic chemistry kits Nos. 6949E, 6949G-2 and 6949K.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) For bleeding gases, steam or liquids
Includes Capillary tip
Length (English) 11 in.
Length (Metric) 280 mm
Material Borosilicate Glass
Product Type Bleed Glass Joint
WARNING: Reproductive Harm - www.P65Warnings.ca.gov