Learn More
Cole-Parmer Pyrex 3140-100 Brand 3140 dish; 100 x 50 mm, pack of 6
Supplier:  Cole-Parmer 3455004
Encompass_Preferred
The rims on these dishes are reinforced and fire polished to reduce chipping. Dishes withstand sterilization(wet or dry). Ideal for storage and crystallization.
Catalog No. NC9950024