Learn More
Corning™ PYREX™ Distilling Adapter Tube with Side Arm and standard taper Joints
GREENER_CHOICE
Catalog No. 0103550 Shop All Corning Products
Change view
Click to view available options
Standard Taper Joint:
24/20
Catalog No. Standard Taper Joint
01-035-50 24/20
1 options
This item is not returnable. View return policy
Catalog No. 01-035-50 Supplier Corning™ Supplier No. 894624
Add to Cart
Add to Cart
This item is not returnable. View return policy
Adapter with standard taper 24/40 outer joint at top and standard taper 24/40 inner joint at bottom
These PYREX™ 105° angle vacuum connecting adapters have an outer 24/40 Standard Taper joint at the top and an inner-sealed through drip-tip joint at the bottom. Bent at a 105° angle. The serrated sidearm has a 10mm O.D. Drip-tip at the bottom extends about 20mm below the lower end of the joint.
TRUSTED_SUSTAINABILITY
Specifications
Type Connecting Adapter
Standard Taper Joint 24/20
WARNING: Cancer - www.P65Warnings.ca.gov